MOXI FLOW SYSTEM CHECK BEADS (3 VIALS) 1 * 3 items
Catalog # ORFLMXA016-3
Supplier: Orflo
Frequently Bought Together
Ethanol absolute, EMPROVE® EXPERT Ph. Eur., BP, ChP, JP, USP suitable for use as excipient, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Dimethyl sulphoxide, EMSURE® ACS for analysis, Supelco®
Supplier: Merck
Buffers for Biacore Systems
Supplier: Cytiva
Bis(2-butoxyethyl)ether for synthesis, Sigma-Aldrich®
Supplier: Merck
Acetonitrile, ChromAR® for HPLC, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR® Square Polycarbonate Media Bottles
Supplier: VWR Collection
N-Hydroxysuccinimide for synthesis, Sigma-Aldrich®
Supplier: Merck
2-Propanol ≥70% (v/v), SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
11 components: Ca 2mg/l Zr 5mg/l K 5mg/l Zn 5mg/l Na 5mg/l Fe 5mg/l Al 5mg/l Ti 5mg/l Si 5mg/l P 10mg/l Mn 5mg/l in HNO3 5% 1 * 1 L
Supplier: CUSTOM MADE CHEMICALS LAB
SENSOPLATE 24W GLASS BOTTOM W/O LID ST 1 * 12 items
Supplier: Greiner Bio-One
Tetrahydrofuran ≥99.9% inhibitor-free, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
1,2-Dichloroethane ≥99%
Supplier: Thermo Fisher Scientific
Dichloromethane 99.8% stabilised for HPLC
Supplier: Thermo Fisher Scientific
Acetone ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Scoops for foodstuffs, SteriPlast®
Supplier: BURKLE
Ethanol absolute for histology, J.T.Baker®
Supplier: Avantor