300 g Hexamethylene tetramine réf VWR 24560 22,8 ml Ammoniaque 25 % réf MERCK105432 QSP 1000 eau désionisé 1 * 1 L
Catalog # PRLS87298.290
Supplier: PROLABO CMP
300 g Hexamethylene tetramine réf VWR 24560 22,8 ml Ammoniaque 25 % réf MERCK105432 QSP 1000 eau désionisé 1 * 1 L
Catalog # PRLS87298.290
Supplier: PROLABO CMP
Frequently Bought Together
Test strips for peracetic acid, MQuant®, Supelco®
Supplier: Merck
Test Strips, nitrate, MQuant®, Supelco®
Supplier: Merck
Ammonia solution 25%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Visual colour kits, total hardness, MQuant®, Supelco®
Supplier: Merck
Sodium hydroxide, pellets, EMSURE® for analysis, Supelco®
Supplier: Merck
VWR®, Immersion Oil
Supplier: VWR Collection
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals
Test strips for peroxide, MQuant®, Supelco®
Supplier: Merck
Test strips for ammonium, MQuant®, Supelco®
Supplier: Merck
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Sodium hydroxide 98.5-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
VWR® Rota®, pH indicator paper, 1 to 14
Supplier: VWR Chemicals
Schiff reagent for histology, Sigma-Aldrich®
Supplier: Merck
Test kits, COD cell test, Spectroquant®, Supelco®
Supplier: Merck
Test kits, surfactants, Spectroquant®, Supelco®
Supplier: Merck