Instrument Calibration Standard 1 - 4 components: 5000mg/l each of Ca 1 * 500 mL
Catalog # CMPLN9300218.L5
Supplier: CUSTOM MADE CHEMICALS LAB
Instrument Calibration Standard 1 - 4 components: 5000mg/l each of Ca 1 * 500 mL
Catalog # CMPLN9300218.L5
Supplier: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
Sodium hydroxide 50% in aqueous solution, TECHNICAL
Supplier: VWR Chemicals
Visual colour kits, total hardness, MQuant®, Supelco®
Supplier: Merck
Ammonia solution 25%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Sodium hydroxide, pellets, EMSURE® for analysis, Supelco®
Supplier: Merck
Test strips for free chlorine, MQuant®, Supelco®
Supplier: Merck
Sodium hydroxide 10 mol/l, EMPROVE® EXPERT for biopharmaceutical production, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Sodium hydroxide 98.5-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
pH indicator papers, MQuant®, Supelco®
Supplier: Merck
Sodium hypochlorite (12% Cl₂) in aqueous solution, GPR RECTAPUR®
Supplier: VWR Chemicals
Sodium hydroxide ≥97.0%, shot, TECHNICAL
Supplier: VWR Chemicals
Formic acid ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Sodium hydroxide, pellets, pure, Supelco®
Supplier: Merck
Formalin 10%, Q Path®, buffered
Supplier: VWR Chemicals
Fast Red RC Salt, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Test kits, photometric, COD, LCK cuvette tests
Supplier: Hach