ContactUsLinkComponent
8 components: Dextran 40 [CAS:9004-54-0] 5g/l D-(+)-Maltose Monohydrate [CAS:6363-53-7] 6g/l Maltotriose [CAS:1109-28-0] 6g/l D-(+)-Glucose [CAS:50-99-7] 10g/l L-(+)-Lactic acid [CAS:79-33-4] 5g/l Acetic acid [CAS:64-19-7] 1g/l Ethanol [C 1 * 10 items
  CMPLRD10002810.10
8 components: Dextran 40 [CAS:9004-54-0] 5g/l D-(+)-Maltose Monohydrate [CAS:6363-53-7] 6g/l Maltotriose [CAS:1109-28-0] 6g/l D-(+)-Glucose [CAS:50-99-7] 10g/l L-(+)-Lactic acid [CAS:79-33-4] 5g/l Acetic acid [CAS:64-19-7] 1g/l Ethanol [C 1 * 10 items
  CMPLRD10002810.10
Frequently Bought Together