ContactUsLinkComponent
Furnace ashing lv3/11/c550 tMax1100 with FlaP Door anD over-teMPerature liMiter anD therMocouPle hole in the Furnace Door 1 * 1 items
Catalog # NABELV035K11NOTZ1
Supplier:  NABERTHERM
Furnace ashing lv3/11/c550 tMax1100 with FlaP Door anD over-teMPerature liMiter anD therMocouPle hole in the Furnace Door 1 * 1 items
Catalog # NABELV035K11NOTZ1
Supplier:  NABERTHERM
Frequently Bought Together