ContactUsLinkComponent
Furnace MuFFle lt9/11/skM/c550 tMax1100 with with liFt Door Door anD over-teMPerature liMiter anD therMocouPle hole in the rear wall 1 * 1 items
  NABELM095H1ANOTZ
Furnace MuFFle lt9/11/skM/c550 tMax1100 with with liFt Door Door anD over-teMPerature liMiter anD therMocouPle hole in the rear wall 1 * 1 items
  NABELM095H1ANOTZ
Frequently Bought Together