DEXTROSE ANHYDROUS USP EP BP JP 1 * 100 lb
SPCMDE140-100LBBL
Frequently Bought Together
Water USP, JP, Ph. Eur., purified
: VWR Chemicals
Ethyl acetate ≥99%, GPR RECTAPUR®
: VWR Chemicals
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
: VWR Chemicals
Cyclohexane, EMPLURA®, Supelco®
: Merck
Ethanol 70% ≥70% (v/v), TechniSolv®
: VWR Chemicals
Ethanol, GPR RECTAPUR®
: VWR Chemicals
N,N-Dimethylformamide ≥99.5%, GPR RECTAPUR®
: VWR Chemicals