Squeegees, Plastic
Plastic squeegee with foam blade, Green, 610 mm
MICRSQDG-24
These plastic, double foam squeegees are easy to sanitise.
- Will not rust
- Extremely durable
- Synthetic rubber handle with PP frame
- For use with HACCP compliance programs and colour coding
Integrated blade eliminates seams and crevices, reducing places for bacteria to grow. Frame handle is threaded for use with STA series of plastic handles.
- Colour:Green
- Description:Plastic squeegee with foam blade
- Length:610 mm
- Seq:01
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



