Diamond Hydride 2.o, 2.2um, 2.1mm ID x 50mm Length, 120A, column starter set: HPLC Column 1 * 1 items
Catalog # MICS70200-05D-2
Supplier: MICROSOLV
Diamond Hydride 2.o, 2.2um, 2.1mm ID x 50mm Length, 120A, column starter set: HPLC Column 1 * 1 items
Catalog # MICS70200-05D-2
Supplier: MICROSOLV
Frequently Bought Together
Torus 1-AA Column, 130Å, 5 µm, 4.6 mm X 50 mm 1 * 1 items
Supplier: WATERS
Accessories for valves
Supplier: Agilent
*Only for UCD* 1 * 100 items
Supplier: WATERS
Solid phase extraction cartridges, Bond Elut™ SCX
Supplier: VARIAN
L-MARK® LC Autosampler Syringes
Supplier: Trajan Scientific and Medical
GC columns, J&W GS-Alumina
Supplier: Agilent
b.safe Funnels 180
Supplier: Bohlender
Magnesium standard solution, for ICP-MS, Pure Plus grade
Supplier: PerkinElmer
VWR® Hichrom Organic Acid, HPLC Columns
Supplier: Avantor
HPLC columns, Accucore™ RP-MS
Supplier: Thermo Fisher Scientific
Accessories for sample concentrators, Reacti Vap™
Supplier: Thermo Fisher Scientific
UHPLC precolumn filters, UltraShield
Supplier: Restek