Orders LinkContactUsLinkComponent
BucketBinders™ Multi Bucket Systems
BucketBinders™ Multi Bucket Systems
Double bucket system, 2 PP buckets (green/red), 1 set of BucketBinder™ connectors
Catalog # MICRBB-DBL-DGRE
Supplier:   MICRONOVA
BucketBinders™ Multi Bucket Systems
Catalog # MICRBB-DBL-DGRE
Supplier:  MICRONOVA
BucketBinders™ Multi Bucket Systems
BucketBinders™ Multi Bucket Systems
Double bucket system, 2 PP buckets (green/red), 1 set of BucketBinder™ connectors
Price
866,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
BucketBinders™ bucket system is a cost effective, ergonomic and simple that replaces the need for bulky carts.
  • Easy clip action allows attachment before or after bucket is filled with disinfectant
  • Polyketone hooks firmly secure two or three rectangular buckets together
  • Stainless steel base plate attaches at caster for support
  • Sturdy enough to secure two or three buckets and easy to maneuver with up to 15 litres of liquid in each bucket
  • Lightweight; less than 1 kg per pair
  • Allows scale up or down as required
  • Autoclavable
The system offers easy movement, cleaning, and storage. Works well in both small and large controlled environments and the sterile core.
Will house W-94 flat mop Megawringer™, W-9, W-92 flat wringers or W-9B BoxWringer for string mops. Complements all the Micronova M-Zone FlatMops™, SnapMops™ and SlimLine™ mops.
Accessories information: B-7 polypropylene buckets with autoclavable castors available in five colours, (blue, green, red, green and grey) are available as replacement buckets.
Packaging: BucketBinder™ kits include two or three polypropylene buckets with castors (22 L capacity), and one or two sets of BucketBinder bucket connectors.
Specifications
  • Description:
    Double bucket system, 2 PP buckets (green/red), 1 set of BucketBinder™ connectors
  • Seq:
    02
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • BucketBinder, includes set of stainless steel and polyketone connectors
    BucketBinder, includes set of stainless steel and polyketone connectors
    MICRBB-01
    Log in for availability
    1 * 1 SET - 425,00 €