ContactUsLinkComponent
Moticam 1080n int Intermediate Camera for Motic ba series microscope 1/2,8 Still Resolution: 6mp (3264x1836) Live Resolution: fullhd (1920x1080)LCD 11,6 Full HD Motic Images pl 1 * 1 items
Catalog # MOTI1100600101681
Supplier:  MOTIC
undefined
Moticam 1080n int Intermediate Camera for Motic ba series microscope 1/2,8 Still Resolution: 6mp (3264x1836) Live Resolution: fullhd (1920x1080)LCD 11,6 Full HD Motic Images pl 1 * 1 items
Catalog # MOTI1100600101681
Supplier:  MOTIC
Frequently Bought Together