ContactUsLinkComponent
under table shelf that is compatible with 24 x 24 inch optical breadboard and I-125 isolator frame. It clips onto the cross bracket of the Isostation frame and provides a large shelf depth, while still giving leg clearance. The surface provides 200 l 1 * 1 items
Catalog # ECHLNCTS2424
Supplier:  DISCOVER ECHO
under table shelf that is compatible with 24 x 24 inch optical breadboard and I-125 isolator frame. It clips onto the cross bracket of the Isostation frame and provides a large shelf depth, while still giving leg clearance. The surface provides 200 l 1 * 1 items
Catalog # ECHLNCTS2424
Supplier:  DISCOVER ECHO
Frequently Bought Together