ContactUsLinkComponent
Mixer Mill MM500 control for 200 - 230 V, 50/60 Hz, with quick release clamp, without grinding jars and balls. CE conforming and supplied with 24 months warranty, Operating Manual and 2 metre power cable 1 * 1 items
Catalog # RETS20.767.0001
Supplier:  Retsch GmbH
Mixer Mill MM500 control for 200 - 230 V, 50/60 Hz, with quick release clamp, without grinding jars and balls. CE conforming and supplied with 24 months warranty, Operating Manual and 2 metre power cable 1 * 1 items
Catalog # RETS20.767.0001
Supplier:  Retsch GmbH
Frequently Bought Together