Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
100mg/l each of As,Be,Bi,Ca,Cd,Co,Cr,Cu,Fe,Li,Mg,Mn,Mo,Ni,Pb,Sb,Se,Sr,Ti,Tl,V,Zn 1 * 500 mL
Catalog # CMPLM52B5.K1.5N.L5
Supplier:
CUSTOM MADE CHEMICALS LAB
Expand image
100mg/l each of As,Be,Bi,Ca,Cd,Co,Cr,Cu,Fe,Li,Mg,Mn,Mo,Ni,Pb,Sb,Se,Sr,Ti,Tl,V,Zn 1 * 500 mL
Catalog # CMPLM52B5.K1.5N.L5
Supplier:
CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
pH buffers, ready to use
Supplier: WTW
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Ammonium standard solution, 1000 mg/L NH₄ in water (from NH₄Cl) Certipur® standard for Ion Chromatography, Supelco®
Supplier: Merck
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Nitrate standard solution, 1000 mg/L NO₃<sup>-</sup> in Water (from NaNO₃) Certipur®, Supelco®
Supplier: Merck
Potassium hydrogen phthalate, Certipur® Reag. Ph. Eur., Reag. USP volumetric standard, secondary reference material for alkalimetry, Supelco®
Supplier: Merck
Test Strips, nitrate, MQuant®, Supelco®
Supplier: Merck
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Buffer solutions, ready to use, referenced at 20 °C, Certipur®, Supelco®
Supplier: Merck
Potassium chloride 3 mol/l (3 N) in aqueous solution, Supelco®
Supplier: Merck