ContactUsLinkComponent
Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 mg, max. load 61 g. Draft Shield D 1 * 1 items
Catalog # SARBMCA66S3CF123
Supplier:  Sartorius Balances
Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 mg, max. load 61 g. Draft Shield D 1 * 1 items
Catalog # SARBMCA66S3CF123
Supplier:  Sartorius Balances
Frequently Bought Together