Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 mg, max. load 32 g. Draft Shield D & Activated Ionizer 1 * 1 items
Catalog # SARBMCA36S3S0023I
Supplier: Sartorius Balances
Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 mg, max. load 32 g. Draft Shield D & Activated Ionizer 1 * 1 items
Catalog # SARBMCA36S3S0023I
Supplier: Sartorius Balances
Frequently Bought Together
Gas cartridges, Campingaz®
Supplier: CAMPING GAZ
Bunsen burners, LABOGAZ 206
Supplier: CAMPING GAZ
VWR®, Mini vortex mixer
Supplier: VWR Collection
Magnetic hotplate stirrer, RH basic 2 IKAMAG®
Supplier: IKA
Accessories for orbital shakers
Supplier: IKA
Accessories for tube mill control
Supplier: IKA
Magnetic stirrers, MST and MST Digital
Supplier: VELP SCIENTIFIC
Hei-PLATE Mix 'n' Heat Core Magnetic Stirrers
Supplier: Heidolph Instruments GmbH & Co.KG
MR Hei-Mix S Magnetic Stirrer
Supplier: Heidolph Instruments GmbH & Co.KG
Vortex mixer, VORTEX Genius 3
Supplier: IKA
Gas cartridge, CG1750
Supplier: CAMPING GAZ
Accessories for vortex mixers, MS 3
Supplier: IKA
Cartridge 220G for cartridge burner
Supplier: USBECK Laborgeräte
VWR® VMS Advanced Magnetic Hotplate Stirrers
Supplier: VWR Collection
VWR® VWB2, Water Baths
Supplier: VWR Collection
VWR® INCU-Line® IL 10, IL 23, IL 56, Digital Incubators
Supplier: VWR Collection
VWR® VMS-A, Magnetic Hotplate Stirrer
Supplier: VWR Collection
Gas burner, powerjet/powerjet 2
Supplier: WARTEWIG
Magnetic stirring bars
Supplier: VELP SCIENTIFIC