ContactUsLinkComponent
SpectraMax PARADIGM Tunable (TUNE) Detection Cartridge is optimized for 96-, 384-, and 1536-well plates and provides wavelength sele ction in 1nm increments in Fluorescence Intensity (FI),Time Resolved Fluorescence (TRF), and Luminescence (LUM) detec 1 * 1 items
Catalog # MLDV0200-7050-TR
Supplier:  Molecular Devices
SpectraMax PARADIGM Tunable (TUNE) Detection Cartridge is optimized for 96-, 384-, and 1536-well plates and provides wavelength sele ction in 1nm increments in Fluorescence Intensity (FI),Time Resolved Fluorescence (TRF), and Luminescence (LUM) detec 1 * 1 items
Catalog # MLDV0200-7050-TR
Supplier:  Molecular Devices
Frequently Bought Together