ContactUsLinkComponent
SpectraMax AlphaScreen 1536 HTS Detection Cartidge utilize a 680 nm Laser Diode to provide a sensitive reading system for AlphaScree n assays. Compatible with SpectraMax PARADIGM and SpectraMax i3x systems. In addition, a patent pending design that i 1 * 1 items
Catalog # MLDV0200-7019-TR
Supplier:  Molecular Devices
SpectraMax AlphaScreen 1536 HTS Detection Cartidge utilize a 680 nm Laser Diode to provide a sensitive reading system for AlphaScree n assays. Compatible with SpectraMax PARADIGM and SpectraMax i3x systems. In addition, a patent pending design that i 1 * 1 items
Catalog # MLDV0200-7019-TR
Supplier:  Molecular Devices
Frequently Bought Together