ContactUsLinkComponent
SpectraMax Glow Luminescence (LUM) (384) Detection Cartridge for 384-well plates, utilizes a unique, well-isolating design that sign ificantly reduces the level of cross talk from surrounding wells. Compatible with SpectraMax PARADIGM and/or Spectra 1 * 1 items
Catalog # MLDV0200-7015-TR
Supplier:  Molecular Devices
SpectraMax Glow Luminescence (LUM) (384) Detection Cartridge for 384-well plates, utilizes a unique, well-isolating design that sign ificantly reduces the level of cross talk from surrounding wells. Compatible with SpectraMax PARADIGM and/or Spectra 1 * 1 items
Catalog # MLDV0200-7015-TR
Supplier:  Molecular Devices
Frequently Bought Together