ContactUsLinkComponent
SpectraMax Glow Luminescence (LUM) Detection Cartridge optimized for 96, 384 and 1536-well plates, utilizes a unique, well-isolating design that significantly reduces the level of cross talk from surrounding wells. Compatible with SpectraMax PARADIG 1 * 1 items
Catalog # MLDV0200-7012-TR
Supplier:  Molecular Devices
SpectraMax Glow Luminescence (LUM) Detection Cartridge optimized for 96, 384 and 1536-well plates, utilizes a unique, well-isolating design that significantly reduces the level of cross talk from surrounding wells. Compatible with SpectraMax PARADIG 1 * 1 items
Catalog # MLDV0200-7012-TR
Supplier:  Molecular Devices
Frequently Bought Together