ContactUsLinkComponent
SpectraMax Cisbio HTRF Detection Cartridge utilizes a high energy Xenon flash lamp for sensitive reading of HTRF reagents. Compatib le with SpectraMax PARADIGM and/or SpectraMax i3/i3X systems. In addition, the standard dual emission design gives th 1 * 1 items
Catalog # MLDV0200-7011-TR
Supplier:  Molecular Devices
SpectraMax Cisbio HTRF Detection Cartridge utilizes a high energy Xenon flash lamp for sensitive reading of HTRF reagents. Compatib le with SpectraMax PARADIGM and/or SpectraMax i3/i3X systems. In addition, the standard dual emission design gives th 1 * 1 items
Catalog # MLDV0200-7011-TR
Supplier:  Molecular Devices
Frequently Bought Together