Square HDPE Bottle
1000 ml, Square bottle, HDPE
SAMP8165A-1000
Square HDPE bottles for use with a range of liquids and powders.
- Different sizes available
- Ribbed sides for easy handling
- Available from stock
- Fast delivery
This range of HDPE containers are perfect for use with a range of powders and liquids. They have a handy square space-saving shape and incorporate side ridges for easy handling when wet.
- Capacity:1000 ml
- Description:Square bottle
- Material:HDPE
- Seq:03