R-PLEX Human PF-4/CXCL4 Antibody Set (5 Plate Size) 1 * 5 items
Catalog # MESOF21R1-3
Supplier: MESO SCALE DISCOVERY
R-PLEX Human PF-4/CXCL4 Antibody Set (5 Plate Size) 1 * 5 items
Catalog # MESOF21R1-3
Supplier: MESO SCALE DISCOVERY
Frequently Bought Together
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
Individually Wrapped Serological Pipettes, Falcon®
Supplier: Corning
VWR® Inoculating Loops and Needles, Disposable
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
Round-bottom polystyrene tubes, disposable, Falcon®
Supplier: Corning
Reagent/centrifuge tube with screw cap
Supplier: Greiner Bio-One
Eppendorf® DNA LoBind® Microcentrifuge Tubes
Supplier: EPPENDORF
Serological pipettes, Stripette™
Supplier: Corning
Cell culture flasks, Nunc™
Supplier: Thermo Fisher Scientific
Eppendorf® Safe-Lock Tubes, 0,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection
Eppendorf Tubes® Microtubes with Snap Cap, 5,0 ml
Supplier: EPPENDORF
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
R-PLEX Human Serpin E1 (total) Antibody Set 1 * 5 items
Supplier: MESO SCALE DISCOVERY