0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Belgium
ContactUsLinkComponent
21-(Cyclohexylidene)pregn-4-ene-3,20-dione pharmaceutical secondary standard, certified reference material 1 * 100 mg
PHR8940-100MG
:
Merck
21-(Cyclohexylidene)pregn-4-ene-3,20-dione pharmaceutical secondary standard, certified reference material 1 * 100 mg
PHR8940-100MG
:
Merck
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
: Merck
VWR® Buffer Solutions, Secondary Standards
: VWR Chemicals
Buffer solutions, ready to use, referenced at 20 °C, Certipur®, Supelco®
: Merck
Ammonium standard solution, 1000 mg/L NH₄ in water (from NH₄Cl) Certipur® standard for Ion Chromatography, Supelco®
: Merck
Buffer solutions, ready to use, referenced at 25 °C, Certipur®
: Merck
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
: VWR Chemicals
VWR®, Conductivity standard solutions
: VWR Chemicals
pH buffers, ready to use
: WTW
Conductivity standards, KCl, referenced at 25 °C, for conductivity measurement
: Mettler - Toledo