Fluorometholone certified reference material, pharmaceutical secondary standard 1 * 400 mg
Catalog # PHR3405-400MG
Supplier: Merck
Fluorometholone certified reference material, pharmaceutical secondary standard 1 * 400 mg
Catalog # PHR3405-400MG
Supplier: Merck
Frequently Bought Together
Supelco® COD CRM, standard solutions
Supplier: Merck
VWR®, Viscosity standards
Supplier: VWR Chemicals
pH buffer solutions
Supplier: Mettler - Toledo
Calcium 1.000 mg/l in water standard for ion chromatography
Supplier: VWR Chemicals
VWR®, Chlorite 1,000 mg/L in NaOH solution standard for ion chromatography
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
pH buffer solutions, certified (DKD)
Supplier: Mettler - Toledo
N-Nitroso-ethyl-isopropylamine 100 µg/mL in Methanol 1 * 1 mL
Supplier: EHRENSTORFER
Turbidity standards, Stablcal®
Supplier: Hach
Reagents for chloride determination using Lovibond® MD photometers
Supplier: Lovibond
Reagents for alkalinity determination using Lovibond® MD photometers
Supplier: Lovibond
VWR®, Flow Cup viscosity standards
Supplier: VWR Chemicals