Ag Al As B Ba Be Bi Ca Cd Co Cr Cu Fe K Li Mg Mn Mo Na Ni Pb Sb Se Sr Ti Tl V Zn 1 * 500 mL
Catalog # CMPLRM031071L5
Fournisseur: CUSTOM MADE CHEMICALS LAB
Ag Al As B Ba Be Bi Ca Cd Co Cr Cu Fe K Li Mg Mn Mo Na Ni Pb Sb Se Sr Ti Tl V Zn 1 * 500 mL
Catalog # CMPLRM031071L5
Fournisseur: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
Potassium pyroantimonate 0.05 mol/l in potassium hydroxide solution Reag. Ph. Eur. 1071301
Fournisseur: VWR Chemicals
BUFFER PH 4,01 DOSING BOTTLE 1 * 500 mL
Fournisseur: HAMILTON BONADUZ
pH buffers, ready to use
Fournisseur: WTW
Buffer solutions as per DIN 19266-01: 2000
Fournisseur: WTW
VWR®, Redox (ORP) Standards
Fournisseur: VWR Chemicals
pH buffer solutions
Fournisseur: Mettler - Toledo
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Fournisseur: VWR Chemicals
VWR®, Primary colour solutions, Reagent Ph. Eur.
Fournisseur: VWR Chemicals
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Fournisseur: VWR Chemicals
pH buffers, DuraCal
Fournisseur: HAMILTON BONADUZ
Nitrogen (total) CRM, Standard Solutions, Supelco®
Fournisseur: Merck
Titanium trichloride sulphuric acid reagent Reag. Ph. Eur. 1091202
Fournisseur: VWR Chemicals
Potassium chloride 3 mol/l (3 N) in aqueous solution, Supelco®
Fournisseur: Merck
Buffer solutions, ready to use, referenced at 20 °C, Certipur®, Supelco®
Fournisseur: Merck
pH buffer solutions, certified (DKD)
Fournisseur: Mettler - Toledo