Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Accessories for Toggle Drums

Drum lid, 316L SS, 395 mm int.Ø of drum
Catalog # SAMPA429-395
Supplier:   Sampling Systems
Accessories for Toggle Drums
Catalog # SAMPA429-395
Supplier:  Sampling Systems
Accessories for Toggle Drums

Drum lid, 316L SS, 395 mm int.Ø of drum

Price
353,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    Drum lid, 316L SS, 395 mm int.Ø of drum
  • Seq:
    01