AMMONIUM PHOSPHATE MONOBASIC CRYSTAL 1 * 500 g
Catalog # SPCMA1235-500GM
Supplier: Spectrum Chemical
AMMONIUM PHOSPHATE MONOBASIC CRYSTAL 1 * 500 g
Catalog # SPCMA1235-500GM
Supplier: Spectrum Chemical
Frequently Bought Together
Ammonium formate, (max. 3% H₂O) 97%
Supplier: Thermo Fisher Scientific
Cobalt(Ii) Chloride Hexahydrate, 98+% 1 * 1 kg
Supplier: Strem Chemicals, Inc.
Sodium Acetate 3-hydrate for analysis, ACS, ISO 1 * 1.000 g
Supplier: PanReac AppliChem
Lithium chloride 99+% ACS
Supplier: Thermo Fisher Scientific
Silver Nanoparticles (60Nm, 0.02Mg/Ml In 1 * 100 mL
Supplier: Strem Chemicals, Inc.
Titanium Carbide, 99% 1 * 500 g
Supplier: Strem Chemicals, Inc.
Europium(III) chloride hydrate 99,99%
Supplier: Apollo Scientific
Gold Nanoparticles (400Nm Diameter, 1 Od 1 * 25 mL
Supplier: Strem Chemicals, Inc.
Silver Nitrate (99.9%-Ag) (Acs) 1 * 500 g
Supplier: Strem Chemicals, Inc.
1-Pentanesulphonic acid sodium salt monohydrate 98+% for HPLC
Supplier: Thermo Fisher Scientific
Disodium hydrogen phosphate dihydrate, meets analytical specification of Ph. Eur., USP
Supplier: Thermo Fisher Scientific
AMMONIA AS NH3 - 1000 MG/L 1 * 125 mL
Supplier: WATERS
Lead(II) acetate basic ACS
Supplier: Thermo Fisher Scientific