SICHERUNG 5X20MM TRAEGE NR 19181/10A CV 1 * 1 items
Catalog # CERT1230363
Supplier: CERTOCLAV
Frequently Bought Together
Cable set 230V 1 * 1 items
Supplier: MEMMERT
SERVICE KIT PCRW01 HEPA FAN EU/UK 1 * 1 items
Supplier: VWR Collection
Fuse 2 A 230 V 1 * 1 items
Supplier: ELMA SCHMIDBAUER
Water PAN (ILCO) 1 * 1 items
Supplier: VWR Collection
RELAY ASSEMBLY 1 * 1 items
Supplier: Cole Parmer
wooden box for glass door 1 * 1 items
Supplier: Binder
BIOTECTOR BASE REAGENT 1 * 20 L
Supplier: Hach
[EN]VITON O-RINGS F/20CC SS SYR 1 * 10 ST
Supplier: HARVARD APPARATUS
Assembled eccentric group with rev counter 1 * 1 items
Supplier: VELP SCIENTIFIC
Set grad.glass R-6 RV 1ml,6pc 1 * 6 items
Supplier: BUCHI