Orders LinkContactUsLinkComponent
PolyCHECK® Wipes
PolyCHECK® Wipes
Black, 228,6×228,6 mm
Catalog # FOAMHT5291K-5
Supplier:   FOAMTEC
PolyCHECK® Wipes
Catalog # FOAMHT5291K-5
Supplier:  FOAMTEC
PolyCHECK® Wipes
Black, 228,6×228,6 mm
Price
1 790,00 €
Log in for availability
1 * 1.000 ST
Product Details & Documents
PolyCHECK® Wipe, black polyester inspection wipe with sealed edge.
Specifications
  • Colour:
    Black
  • L×W:
    228,6×228,6 mm
  • Seq:
    01