Saisie de commande
Belgium
ContactUsLinkComponent
,,7 components: 5mg/l each of F- Cl- NO2- Br- NO3- PO43- SO42- in H2O ,, ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 12 months. (6 months after opening),, ,,Ambient Temp.,, 1 * 100 mL
Catalog # CMPLRM008475L1
,,7 components: 5mg/l each of F- Cl- NO2- Br- NO3- PO43- SO42- in H2O ,, ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 12 months. (6 months after opening),, ,,Ambient Temp.,, 1 * 100 mL
Catalog # CMPLRM008475L1
Frequently Bought Together