HTD Equilibrium Dialysis Device
Product Details & Documents
HTD reusable equilibrium dialysis device combines industry-leading accuracy and precision with an automation compatible design for a cost efficient, scalable solution.
- Simultaneously process 1 to 96 assays, each with a 30 to 150 µl capacity to conserve valuable reagents
- Compatible with a wide range of dialysis membrane pore sizes
- Ready in minutes, achieves equilibrium in 4 to 6 hours on average
- Automatable with standard robotic workstations and 96-well pipettors
- Ready access to both the sample and dialysate sides of the membrane from the top of the device
- Minimal non-specific binding due to 100% Teflon block construction
- Reusable device is cost-effective, eco-friendly, and scalable
- Compatible with a wide range of dialysis membrane pore sizes: 1 kDa, 3,5 kDa, 6 to 8 kDa, 10 kDa, 12 to 14 kDa, 25 kDa, 50 kDa
The HTD96B micro equilibrium device was developed at a Global Pharmaceutical Research laboratory to assess serum protein binding, microsomal protein binding, and tissue specific binding of drug candidates, conserving sample quantities, and achieving rapid equilibrium.
Reusable device combines industry-leading accuracy and precision with an automation compatible design for a scalable, cost-efficient, and eco-friendly solution.
Applications:
- Serum/plasma protein binding
- Microsome protein binding
- Relative binding of drug to specific tissues
- Receptor binding
- T3/T4 free fraction binding (hyperthyroidism) determinations
The 96C model has a shorter diffusion path with a 25 to 75 µl capacity and achieves equilibrium in 2 to 4 hours on average.
Additional specifications:
Dimensions: 170,1×116,8×38,1 mm
Weight: 0,54 kg
Specifications
- MWCO:3,5 kDa
- Volume:—
- Description:Equilibrium dialysis membrane, regenerated cellulose
- Seq:7
Accessories
Product Information
- Catalog #AvailabilityPrice
- Equilibrium dialysis connecting rods
Product Information
- Catalog #AvailabilityPrice
- 96 well equilibrium dialysis teflon block (shorter diffusion path)96 well equilibrium dialysis teflon block (shorter diffusion path)HTDL1010Log in for availability1 * 1 ST
Product Information
- Catalog #AvailabilityPrice
- Stainless steel springs
Product Information
- Catalog #AvailabilityPrice
- 96 well equilibrium dialysis Teflon block
Product Information
- Catalog #AvailabilityPrice
- Equilibrium dialysis base clamp
Product Information
- Catalog #AvailabilityPrice
- Stainless steel pressure plate
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



