SULFITES 10 MG/L IN H2O 1 * 100 mL
CMPLRM096560L1
Frequently Bought Together
UltraClear® for histology, J.T.Baker®
: Avantor
VWR® Dosatest, pH indicator test strips, non bleeding
: VWR Chemicals
Sodium hydroxide ≥97.0%, shot, TECHNICAL
: VWR Chemicals
Formalin 10%, Q Path®, buffered
: VWR Chemicals
Formaldehyde 36% in aqueous solution stabilised, GPR RECTAPUR®
: VWR Chemicals
pH indicator test strips, pH-Fix
: MACHEREY-NAGEL
Ammonium formate ≥99%, HiPerSolv CHROMANORM® for LC-MS
: VWR Chemicals
Formic acid ≥99%, HiPerSolv CHROMANORM® for LC-MS
: VWR Chemicals