Double Ended Spoons
Double ended spoon, 180 mm, 0,8/2,8 ml, Stainless steel
Catalogus # SAMPA104-180
Leverancier: Sampling Systems
Ga naar:
Product Details & Documents
Double ended spoon for taking different sample sizes.
- Made from stainless steel
- Ideal for sampling powders and liquids
- Various sizes available
These double ended spoons are great for taking different sample sizes using the same spoon. The lengths and volume capacities of the spoons vary allowing for flexibility in sampling both powders and liquids.
Specificaties
- Capacity:0,8/2,8 ml
- Description:Double ended spoon
- Length:180 mm
- Material:Stainless steel
- Seq:05
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.