Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
As B Ba Cd Cr Co Cu Hg Mo Mn Ni Pb Sb Se Sn Tl V Zn in HNO3 5% 1 * 100 mL
Catalog # CMPLRM097771L1
Supplier:
CUSTOM MADE CHEMICALS LAB
Expand image
As B Ba Cd Cr Co Cu Hg Mo Mn Ni Pb Sb Se Sn Tl V Zn in HNO3 5% 1 * 100 mL
Catalog # CMPLRM097771L1
Supplier:
CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
Sodium hydroxide, pellets, EMSURE® for analysis, Supelco®
Supplier: Merck
pH indicator papers, MQuant®, Supelco®
Supplier: Merck
Supelco® Spectroquant® Test Kits, Sulphate
Supplier: Merck
Test kits, nitrite, Spectroquant®, Supelco®
Supplier: Merck
Test kits, nitrogen (total) cell tests, Spectroquant®, Supelco®
Supplier: Merck
Test kits, COD cell test, Spectroquant®, Supelco®
Supplier: Merck
Giemsa's stain solution for microscopy, Sigma-Aldrich®
Supplier: Merck
Test kits, surfactants, Spectroquant®, Supelco®
Supplier: Merck
Test strips for peroxide, MQuant®, Supelco®
Supplier: Merck
Test kits, ammonium, Spectroquant®, Supelco®
Supplier: Merck
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Aqueous solution containing sulphuric acid (0.001M) and Triton X-100 (0.225% w/v) 1 * 10 L
Supplier: CUSTOM MADE CHEMICALS
Test kits, photometric, Nitrate, LCK cuvette tests
Supplier: Hach