ditert-butyl (2R,5R)-5-hydroxy-5-methyl-piperidine-1,2-dicarboxylate 1 * 1 g
Catalog # PHBLPBZT380-1G
Supplier: Pharmablock (USA)
ditert-butyl (2R,5R)-5-hydroxy-5-methyl-piperidine-1,2-dicarboxylate 1 * 1 g
Catalog # PHBLPBZT380-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Ethanol absolute ≥99.5%, EMPLURA®, Supelco®
Supplier: Merck
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Toluene, anhydrous (max. 0.005% H₂O) ≥99.8%, AnalaR NORMAPUR® for synthesis
Supplier: VWR Chemicals
Petroleum spirit, 40…60 °C, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Ethanol absolute ≥99.5%, TechniSolv®, pure
Supplier: VWR Chemicals
N,N-Dimethylformamide ≥99.9%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals