{1-[(tert-butoxy)carbonyl]piperidin-4-yl}methyl 4-fluoro-4-(hydroxymethyl)piperidine-1-carboxylate 1 * 1 g
Catalog # PHBLPBZS3142-1G
Supplier: Pharmablock (USA)
{1-[(tert-butoxy)carbonyl]piperidin-4-yl}methyl 4-fluoro-4-(hydroxymethyl)piperidine-1-carboxylate 1 * 1 g
Catalog # PHBLPBZS3142-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
(±)-Tetrahydrofurfuryl alcohol, Sigma-Aldrich®
Supplier: Merck
Dimethyl sulfoxide EMPLURA 1 * 185 L
Supplier: Merck
Acetone ≥99.5%, Laboratory Reagent
Supplier: Honeywell Chemicals
Ethanolamine ACS
Supplier: Thermo Fisher Scientific
Petroleum spirit, 40…60 °C, extra pure
Supplier: Thermo Fisher Scientific
ETHYL ACETATE 99.5% FOR SYNTHESIS 1 * 200 L
Supplier: PanReac AppliChem
Methanol ≥99.9% (by GC) Reag. Ph. Eur., ACS, ISO for analysis
Supplier: PanReac AppliChem
2-Methylbutane 99+%, extra pure
Supplier: Thermo Fisher Scientific
Water, GPR RECTAPUR®
Supplier: VWR Chemicals
Benzonitrile 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
n-Hexane
Supplier: Roth Carl
n-Hexane 95%, ChromAR®, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Diethyl ether ≥99% stabilised for HPLC
Supplier: Thermo Fisher Scientific