tert-butyl 4-[(4-bromo-1H-pyrazol-1-yl)methyl]piperidine-1-carboxylate 1 * 1 g
Catalog # PHBLPBXA1921-1G
Supplier: Pharmablock (USA)
tert-butyl 4-[(4-bromo-1H-pyrazol-1-yl)methyl]piperidine-1-carboxylate 1 * 1 g
Catalog # PHBLPBXA1921-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Chloroform 99.0-99.6% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Xylene (mixture of isomers) ≥98.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol, GPR RECTAPUR®
Supplier: VWR Chemicals
n-Hexane, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
n-Heptane ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
1-Butanol ≥98.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethylene glycol ≥99.7%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Ethyl acetate ≥99%, TECHNICAL
Supplier: VWR Chemicals
Ethyl acetate, EMPROVE® ESSENTIAL Ph. Eur., BP, NF, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
2-Propanol ≥99.0% USP, Ph. Eur.
Supplier: VWR Chemicals