1-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-3-carbonitrile 1 * 1 g
PHBLPBXA1494-1G
1-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-3-carbonitrile 1 * 1 g
PHBLPBXA1494-1G
Frequently Bought Together
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
1-Octanol, EMPLURA®, Supelco®
: Merck
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
: VWR Chemicals
2,2,4-Trimethylpentane ≥99.5%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Petroleum spirit, 30…40 °C, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Dimethyl sulphoxide ≥99.7%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
: Honeywell Chemicals
N,N-Dimethylformamide ≥99.7% for HPLC
: Thermo Fisher Scientific