N-[1-[(2R,3R,4R,5R)-5-[[bis(4-methoxyphenyl)-phenyl-methoxy]methyl]-4-[2-cyanoethoxy-(diisopropylamino)phosphanyl]oxy-3-(2-methoxyet hoxy)tetrahydrofuran-2-yl]-2-oxo-pyrimidin-4-yl]benzamide 1 * 1 g
Catalog # PHBLPBT8307-1G
Supplier: Pharmablock (USA)
N-[1-[(2R,3R,4R,5R)-5-[[bis(4-methoxyphenyl)-phenyl-methoxy]methyl]-4-[2-cyanoethoxy-(diisopropylamino)phosphanyl]oxy-3-(2-methoxyet hoxy)tetrahydrofuran-2-yl]-2-oxo-pyrimidin-4-yl]benzamide 1 * 1 g
Catalog # PHBLPBT8307-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Glycerine 85% Ph. Eur.
Supplier: VWR Chemicals
Toluene ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
2-Propanol ≥99.5% (by GC), EMPLURA®, Supelco®
Supplier: Merck
Tetrahydrofuran ≥99.9% unstabilised, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Ethanol absolute ≥99.5%, TechniSolv®, pure
Supplier: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
Supplier: VWR Chemicals
Ethyl acetate ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Chloroform ≥99.8% stabilised, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethanol 96% ≥96% (v/v), TechniSolv®, pure
Supplier: VWR Chemicals