tert-butyl N-[2-[2-[2-[2-[2-[2-(2-aminoethoxy)ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethyl]carbamate 1 * 1 g
Catalog # PHBLPBT5458-1G
Supplier: Pharmablock (USA)
tert-butyl N-[2-[2-[2-[2-[2-[2-(2-aminoethoxy)ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethyl]carbamate 1 * 1 g
Catalog # PHBLPBT5458-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Dichloromethane ≥98% stabilised, TECHNICAL
Supplier: VWR Chemicals
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Toluene, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
Water, AnalaR NORMAPUR® ISO 3696 Grade 3 analytical reagent
Supplier: VWR Chemicals
Xylene (mixture of isomers) ≥98.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals
Acetonitrile : water : trifluoroacetic acid (50 : 47,5 : 2,5), HiPerSolv CHROMANORM®
Supplier: VWR Chemicals
p-Xylene ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethanol absolute ≥99%
Supplier: KLINIPATH
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Acetone ≥99.8%, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
2-Propanol, EMPROVE® ESSENTIAL Ph. Eur., BP, JP, USP, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Ethyl acetate, anhydrous (max. 0.005% H₂O) ≥99.8%
Supplier: VWR Chemicals
Acetic acid 99-100% for synthesis, Sigma-Aldrich®
Supplier: Merck
Cyclohexane ≥99.8%, SupraSolv® for GC-MS
Supplier: Merck
Glycerine 87% 86%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals