(1R)-2-[(tert-butyldimethylsilyl)oxy]-1-(4-chloro-3-fluorophenyl)ethan-1-ol 1 * 1 g
Catalog # PHBLPBLJ7703-1G
Supplier: Pharmablock (USA)
(1R)-2-[(tert-butyldimethylsilyl)oxy]-1-(4-chloro-3-fluorophenyl)ethan-1-ol 1 * 1 g
Catalog # PHBLPBLJ7703-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Benzyl alcohol ≥99.0% analytical reagent
Supplier: VWR Chemicals
Benzyl acetate for synthesis, Sigma-Aldrich®
Supplier: Merck
Bis(2-methoxyethyl)ether, extra dry over Molecular Sieve 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide, extra dry over Molecular Sieve 99.7+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Ethyl acetate 99.6% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
Chloroform 99.0-99.4% (by GC) stabilised, Puriss. p.a. ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Methanol ≥99.9% (by GC) Reag. Ph. Eur., ACS, ISO for analysis
Supplier: PanReac AppliChem
(±)-2-Methyltetrahydrofuran, extra dry 99+% stabiliser free, AcroSeal®
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide (DMSO),?99,9 %, Ph. Eur., USP 1 * 500 mL
Supplier: Roth Carl
Petroleum spirit, 80…110 °C
Supplier: Roth Carl
Ethanol (untaxed) in water ≥70% (v/v) Ph. Helv., Riedel-de Haën™
Supplier: Honeywell Chemicals
Cyclohexane 99.8% for HPLC
Supplier: Thermo Fisher Scientific