1-methyl-3-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrolo[2,3-b]pyridine 1 * 1 g
Catalog # PHBLPBLJ1168-1G
Supplier: Pharmablock (USA)
1-methyl-3-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrolo[2,3-b]pyridine 1 * 1 g
Catalog # PHBLPBLJ1168-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
(R)-(+)-Pulegone for synthesis, Sigma-Aldrich®
Supplier: Merck
N,N-Dimethylformamide anhydrous, 99.8 % 1 * 1 L
Supplier: Merck
2-Propanol, EMPROVE® ESSENTIAL Ph. Eur., BP, JP, USP, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Petroleum spirit, 40…80 °C, EMPLURA®, Supelco®
Supplier: Merck
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetonitrile ≥99%, TECHNICAL
Supplier: VWR Chemicals
Methanol ≥99.7%, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
N-Methyl-2-pyrrolidone (NMP) 99+% ACS
Supplier: Thermo Fisher Scientific
N,N-Dimethylacetamide ≥99%
Supplier: Thermo Fisher Scientific
Xylene (mixture of isomers) ≥98.5% (isomers plus ethylbenzene) ACS
Supplier: Thermo Fisher Scientific
N,N-Dimethylformamide ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethyl acetate, AR®, Laboratory Reagent, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Benzonitrile for HPLC
Supplier: Thermo Fisher Scientific
2,4,6-Tris(2-pyridyl)-1,3,5-triazine, Supelco®
Supplier: Merck
(±)-2-Methyltetrahydrofuran, anhydrous stabilised for grignard-reaction
Supplier: Thermo Fisher Scientific
N-Methyl-2-pyrrolidone (NMP), anhydrous ≥99.5%
Supplier: Thermo Fisher Scientific