5-azaspiro[3.5]nonan-8-one hydrochloride 1 * 1 g
Catalog # PHBLPBES00554-1-1G
Supplier: Pharmablock (USA)
5-azaspiro[3.5]nonan-8-one hydrochloride 1 * 1 g
Catalog # PHBLPBES00554-1-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Butyl acetate ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
1,3-Propanediol ≥99.7%, GPR RECTAPUR® for synthesis
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99.9%, PESTINORM® for capillary GC analysis
Supplier: VWR Chemicals
Toluene ≥99.90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Tetrahydrofuran ≥99.9% unstabilised, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Ethanol, GPR RECTAPUR®
Supplier: VWR Chemicals
N,N-Dimethylformamide, extra dry over Molecular Sieve 99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific
n-Octane 97%, pure
Supplier: Thermo Fisher Scientific
2-Propanol ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals