trans-1,2-cyclobutanedicarboxylic acid 1 * 5 g
PHBLPB05702-5G
Frequently Bought Together
Xylene - Mixture of isomers technical grade 1 * 5 L
: PanReac AppliChem
2-Methylbutane 99+%, extra pure
: Thermo Fisher Scientific
1,4-Dioxane ≥99% for HPLC
: Thermo Fisher Scientific
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
: VWR Chemicals
Carbon disulphide 99.9% for HPLC
: Thermo Fisher Scientific
Chloroform ≥99.8% stabilised ACS, Riedel-de Haën™
: Honeywell Chemicals
Butanone (Methylethylketone) pure 1 * 2,5 L
: PanReac AppliChem
Benzonitrile 99+% for spectroscopy
: Thermo Fisher Scientific
n-Hexane ≥97%, CHROMASOLV™ for LC-MS, Riedel-de Haën™
: Honeywell Chemicals
2,2,4-Trimethylpentane 99.5% for analysis
: Thermo Fisher Scientific
Polyvinyl alcohol, M.W. ∼ 13,000 98%, hydrolysed
: Thermo Fisher Scientific
ETHYL ACETATE 99.5% FOR SYNTHESIS 1 * 200 L
: PanReac AppliChem