methyl cis-4-hydroxycyclohexanecarboxylate 1 * 5 g
PHBLPB05684-5G
Frequently Bought Together
2-Methylbutane ≥99%, GPR RECTAPUR®
: VWR Chemicals
2-Propanol ≥70% (v/v), SAFC®
: MERCK PRODUCTION CHEMICALS
2,2,4-Trimethylpentane, extra dry over Molecular Sieve 99.5%, AcroSeal®
: Thermo Fisher Scientific
Toluene ≥99.3%, Laboratory Reagent
: Honeywell Chemicals
N,N-Dimethylformamide ≥99.7% for HPLC
: Thermo Fisher Scientific
Methanol ≥99.9%, CHROMASOLV™, gradient grade for HPLC, Riedel-de Haën™
: Honeywell Chemicals
Pyridine ≥99.0% ACS, Riedel-de Haën™
: Honeywell Chemicals
Tetrahydrofuran stabilised ACS, Burdick & Jackson™
: Honeywell Chemicals
Acetone ≥99.8% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
: Honeywell Chemicals
n-Heptane ≥99% (GC (n-heptane basis)), Puriss. p.a. Reag. Ph. Eur., Riedel-de Haën™
: Honeywell Chemicals
Petroleum spirit, 60…80 °C
: Thermo Fisher Scientific
Pyridine ≥99.5% (by GC), Puriss. p.a. ACS, Reag. Ph. Eur., Riedel-de Haën™
: Honeywell Chemicals
Methanol ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
: Honeywell Chemicals
Pyridine 99+% ACS
: Thermo Fisher Scientific