Order Entry
Belgium
Orders LinkContactUsLinkComponent
VWR® Serum/Sample Vials

VWR® Serum/Sample Vials

Neck size: ND20, 20,00 ml, Flat bottom, 24,00×74,00 mm, Clear, No, -
Catalog # 216-3049
Supplier:   VWR Collection
VWR® Serum/Sample Vials
Catalog # 216-3049
Supplier:  VWR Collection
VWR® Serum/Sample Vials
VWR® Serum/Sample Vials

Neck size: ND20, 20,00 ml, Flat bottom, 24,00×74,00 mm, Clear, No, -

Price
55,20 €
Log in for availability
1 * 145 ST
Product Details & Documents

Crimp neck vials with flat bottom made from clear borosilicate glass.

  • Highly resistant against chemicals
  • High temperature resistance
  • Can be used as vaccine or injectable drug containers
  • Stoppers and caps have to be ordered separately

Ideal for long-term sample storage or lyophilisation.

Specifications
  • Cap included:
    No
  • Ø×H:
    24,00×74,00 mm
  • Sterile:
    -
  • Capacity:
    20,00 ml
  • Colour:
    Clear
  • Description:
    Neck size: ND20
  • Base:
    Flat bottom
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.