VWR® Serum/Sample Vials
Neck size: ND20, 5,00 ml, Flat bottom, 24,00×33,00 mm, Clear, No, -
Supplier: VWR Collection
Jump to:
Product Details & Documents
Crimp neck vials with flat bottom made from clear borosilicate glass.
- Highly resistant against chemicals
- High temperature resistance
- Can be used as vaccine or injectable drug containers
- Stoppers and caps have to be ordered separately
Ideal for long-term sample storage or lyophilisation.
Specifications
- Cap included:No
- Ø×H:24,00×33,00 mm
- Sterile:-
- Capacity:5,00 ml
- Colour:Clear
- Description:Neck size: ND20
- Base:Flat bottom
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



