Order Entry
Belgium
Orders LinkContactUsLinkComponent
VWR® Serum/Sample Vials

VWR® Serum/Sample Vials

Neck size: ND13, 3,00 ml, Flat bottom, 16,25×35,00 mm, Clear, No, -
Catalog # 216-3047
Supplier:   VWR Collection
VWR® Serum/Sample Vials
Catalog # 216-3047
Supplier:  VWR Collection
VWR® Serum/Sample Vials
VWR® Serum/Sample Vials

Neck size: ND13, 3,00 ml, Flat bottom, 16,25×35,00 mm, Clear, No, -

Price
30,80 €
Log in for availability
1 * 209 ST
Product Details & Documents

Crimp neck vials with flat bottom made from clear borosilicate glass.

  • Highly resistant against chemicals
  • High temperature resistance
  • Can be used as vaccine or injectable drug containers
  • Stoppers and caps have to be ordered separately

Ideal for long-term sample storage or lyophilisation.

Specifications
  • Cap included:
    No
  • Ø×H:
    16,25×35,00 mm
  • Sterile:
    -
  • Capacity:
    3,00 ml
  • Colour:
    Clear
  • Description:
    Neck size: ND13
  • Base:
    Flat bottom