VWR® Serum/Sample Vials
Neck size: ND11, 2,00 ml, Flat bottom, 10,25×45,00 mm, Clear, No, -
Catalog # 216-3040
Supplier: VWR Collection
Jump to:
Product Details & Documents
Crimp neck vials with flat bottom made from clear borosilicate glass.
- Highly resistant against chemicals
- High temperature resistance
- Can be used as vaccine or injectable drug containers
- Stoppers and caps have to be ordered separately
Ideal for long-term sample storage or lyophilisation.
Specifications
- Cap included:No
- Ø×H:10,25×45,00 mm
- Sterile:-
- Capacity:2,00 ml
- Colour:Clear
- Description:Neck size: ND11
- Base:Flat bottom



